Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine Kinase MM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112842
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112842 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112842 Supplier Thermo Scientific Supplier No. PA5112842
Only null left
Add to Cart
Add to Cart

Specifications

Antigen Creatine Kinase MM
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene CKM
Gene Accession No. P06732, P07310
Gene Alias CKM; CKMM; CPK-M; Creatine kinase M chain; creatine kinase M-type; creatine kinase, M-type; creatine kinase, muscle; creatine phosphokinase M-type; M creatine phosphokinase; MCK; M-CK; muscle creatine kinase
Gene Symbols CKM
Host Species Rabbit
Immunogen Recombinant Human Creatine kinase M-type protein (1-379AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1158, 12715
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.