Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine kinase MT 1A Rabbit anti-Human, Alexa Fluor 700, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB064953 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064953 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064953 Supplier Novus Biologicals Supplier No. NBP300227AF700

Rabbit Polyclonal Antibody

Creatine kinase MT 1A Polyclonal specifically detects Creatine kinase MT 1A in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Creatine kinase MT 1A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias Acidic-Type Mitochondrial Creatine Kinase, CKMT, CKMT1, CKMT1A, Creatine Kinase U-Type, Mitochondrial, Creatine Kinase, Mitochondrial 1 (Ubiquitous), Creatine Kinase, Mitochondrial 1A, EC 2.7.3.2, Mia-CK, Ubiquitous Mitochondrial Creatine Kinase, U-MtCK
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Creatine kinase MT 1A (P12532-1; Ala40-His417).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 548596
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.