Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatine Kinase, Muscle/CKMM Antibody (CKMM/3341), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB381776 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381776 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381776 Supplier Novus Biologicals Supplier No. NBP308304AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Creatine Kinase, Muscle/CKMM Monoclonal antibody specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Creatine Kinase, Muscle/CKMM
Applications ELISA, Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone CKMM/3341
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK
Host Species Mouse
Immunogen Recombinant full-length Creatine Kinase, Muscle/CKMM protein (Uniprot: P06732)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1158
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.