Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Creatinine, NF, 98.5-102%, Spectrum™ Chemical
SDP

Catalog No. 18601872 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
12 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-601-872

Supplier: Spectrum Chemical Manufacturing Corporation C140325KGBL

Only null left
Add to Cart
Add to Cart
Request a Quote

C1403, 60-27-5, C4H7N3O

Spectrum™ Chemical Creatinine, NF - Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 60-27-5
Molecular Formula C4H7N3O
Molecular Weight (g/mol) 113.12
InChI Key DDRJAANPRJIHGJ-UHFFFAOYSA-N
IUPAC Name 2-amino-1-methyl-4,5-dihydro-1H-imidazol-4-one
SMILES CN1CC(=O)N=C1N

Specifications

CAS Max % 100%
Quantity 25 kg
Percent Purity 98.5 to 102%
Grade NF
Ignition Residue 0.2%
Loss on Drying 3%
Packaging Fiber Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.