Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB Antibody (LB9), Novus Biologicals™
SDP

Catalog No. NB10074393 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10074393 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10074393 Supplier Novus Biologicals Supplier No. NB10074393
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

CREB Monoclonal specifically detects CREB in Human, Monkey, Mouse, Primate, Rat samples. It is validated for ChIP Assay, ELISA, IF/IHC, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen CREB
Applications ChIP Assay, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone LB9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, ELISA 0.1 - 1.0 ug/mL, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 2 ug, Immunohistochemistry-Paraffin 1:200, Chromatin Immunoprecipitation (ChIP)
Gene Alias active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Gene Symbols CREB1
Host Species Mouse
Immunogen Recombinant human CREB-A protein.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1385
Test Specificity CREB (LB9)
Target Species Human, Monkey, Rat, Mouse, Primate, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.