Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB, p Ser133 Antibody (10E9), Novus Biologicals™
SDP

Catalog No. NB036575 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036575 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036575 Supplier Novus Biologicals Supplier No. NBP288984
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CREB (p Ser133) Monoclonal antibody specifically detects CREB (p Ser133) in Human, Mouse, Canine samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CREB (p Ser133)
Applications Western Blot, ELISA
Classification Monoclonal
Clone 10000000000
Conjugate Unconjugated
Formulation Lyophilized from 1ml of 2x PBS, PEG, and sucrose
Gene Alias active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Host Species Mouse
Immunogen Synthetic peptide corresponding to a portion of CREBphosphorylated at Ser133.
Purification Method Size Exclusion Chromatography
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 1385
Reconstitution Reconstitute with 1 mL water (15 minutes at room temperature).
Target Species Human, Mouse, Canine
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.