Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB3L2 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB050033 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB050033 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB050033 Supplier Novus Biologicals Supplier No. NBP297960G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CREB3L2 Polyclonal specifically detects CREB3L2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CREB3L2
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias basic transcription factor 2, BBF2 human homolog on chromosome 7, BBF2H7TCAG_1951439, B-ZIB transcription factor, cAMP responsive element binding protein 3-like 2, cAMP-responsive element-binding protein 3-like protein 2, cyclic AMP-responsive element-binding protein 3-like protein 2, FUS/BBF2H7 protein, MGC131709, MGC71006
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human CREB3L2 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64764
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.