Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREB5 Mouse anti-Human, Clone: PCRP-CREB5-1G8, Novus Biologicals™
SDP

Catalog No. NB177593 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177593 20 μg
NB177592 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB177593 Supplier Novus Biologicals Supplier No. NBP31381820UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CREB5 Monoclonal antibody specifically detects CREB5 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Protein Array

Specifications

Antigen CREB5
Applications Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array
Classification Monoclonal
Clone PCRP-CREB5-1G8
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Protein Array
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. Q02930
Gene Alias cAMP response element binding protein CRE-Bpa, cAMP response element-binding protein CRE-BPa, cAMP responsive element binding protein 5, cAMP responsive element binding protein 5 isoform alpha, cAMP-responsive element-binding protein 5, CREB-5, CREBPA, CRE-BPA, cyclic AMP-responsive element-binding protein 5, H_GS165L15.1
Host Species Mouse
Immunogen Recombinant full-length human CREB5 protein (Uniprot: Q02930)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9586
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.