Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREG Antibody (1C6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP242689AH Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242689AH 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242689AH Supplier Novus Biologicals Supplier No. NBP242689AF647

Mouse Monoclonal Antibody

CREG Monoclonal specifically detects CREG in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen CREG
Applications Western Blot, ELISA
Classification Monoclonal
Clone 1C6
Conjugate Alexa Fluor 647
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, protein CREG1
Gene Symbols CREG1
Host Species Mouse
Immunogen Recombinant human CREG1 (32-220aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 8804
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.