Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CREM Antibody, Novus Biologicals™
SDP

Catalog No. NB1001470 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001470 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001470 Supplier Novus Biologicals Supplier No. NB1001470
Add to Cart
Add to Cart

Goat Polyclonal Antibody

CREM Polyclonal specifically detects CREM in Human, Mouse samples. It is validated for Peptide ELISA, Western Blot.

Specifications

Antigen CREM
Applications Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.3 - 1 ug/ml, Peptide ELISA Detection limit 1:16000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_853549
Gene Alias cAMP response element modulator, cAMP responsive element modulator, cAMP-responsive element modulator, hCREM 2alpha-b protein, hCREM 2beta-a protein, hCREM-2, ICER, Inducible cAMP early repressor, inducible cAMP early repressor ICER, MGC111110, MGC17881, MGC41893
Gene Symbols CREM
Host Species Goat
Immunogen Peptide with sequence LETLKDICSPKTDY corresponding to C-Terminus according to NP_853549.1, NP_874387.1, NP_874388.1, NP_874393.1, NP_874394.1, NP_877570.1, NP_877571.1, NP_877572.1, NP_877573.1, NP_878270.1, NP_898883.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1390
Test Specificity This antibody is expected to recognize the following protein isoforms of human CREM protein: a (NP_853549.1); e (NP_874387.1); f (NP_874388.1); k (NP_874393.1); l (NP_874394.1); m (NP_877570.1); n (NP_877571.1); o (NP_877572.1); p (NP_877573.1); q (NP_878270.1); v (NP_898883.1). This antibody is not expected to cross-react with CREB protein.
Target Species Human, Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.