Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRHR1/CRF1 Antibody (343919), Biotin, Novus Biologicals™
SDP

Catalog No. NB371218 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371218 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371218 Supplier Novus Biologicals Supplier No. FAB3930B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRHR1/CRF1 Monoclonal specifically detects CRHR1/CRF1 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen CRHR1/CRF1
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 343919
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias corticotropin releasing hormone receptor 1, corticotropin-releasing factor receptor 1, corticotropin-releasing factor type 1 receptor, Corticotropin-releasing hormone receptor 1, CRF1, CRFR, CRF-R, CRFR1, CRF-R1, CRF-R-1, CRFR-1, CRH-R-1, CRHR1f, CRH-R1h, CRHR1L, CRHRCRH-R1, seven transmembrane helix receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CRHR-1, Ser24-Val415, Accession # NP_004373
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 1394
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.