Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRHR2/CRF2 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056485T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056485T 0.025 mg
NB10056485 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056485T Supplier Novus Biologicals Supplier No. NB10056485SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

CRHR2/CRF2 Polyclonal specifically detects CRHR2/CRF2 in Bovine, Canine, Chicken, Human, Mammalia, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Simple Western, Western Blot.

Specifications

Antigen CRHR2/CRF2
Applications Immunocytochemistry, Immunofluorescence, Simple Western, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Simple Western 2:200
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_001874
Gene Alias corticotropin releasing hormone receptor 2, corticotropin-releasing factor receptor 2, Corticotropin-releasing hormone receptor 2, CRF2, CRF2R, CRFR2, CRF-R2, CRF-R-2, CRFR-2, CRH2R, CRH-R2, CRH-R-2
Gene Symbols CRHR2
Host Species Rabbit
Immunogen A portion of amino acids 75-125 of human CRHR2 was used as the immunogen.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline GPCR, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 1395
Test Specificity There are three isoforms of CRHR2 in human: alpha is 411 amino acids, beta is 438 amino acids, and gamma is 397 amino acids.
Target Species Bovine, Mouse, Canine, Chicken, Human, Chicken, Mammalia, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.