Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIM1 Antibody (MM0202-16G1), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP212207UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212207UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212207UV Supplier Novus Biologicals Supplier No. NBP212207UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRIM1 Monoclonal specifically detects CRIM1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen CRIM1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone MM0202-16G1
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52
Gene Symbols CRIM1
Host Species Mouse
Immunogen Human recombinant CRIM1 (extracellular domain)
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51232
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.