Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ampco Crimped Wire Cup Brush with 6 in. Diameter
SDP

Catalog No. p-200132386
Click to view available options
For Use With (Equipment):
5/8 in. Rotating Shaft
5/8-11 in. Rotating Shaft

Non-sparking, non-magnetic, and corrosion-resistant cup brush.

  • Max RPM: 6,000 RPM

Specifications

Product Type Cup Brush
Diameter (English) 6 in.
For Use With (Equipment) 5/8 in. Rotating Shaft
Quantity 1 Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.