Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIPT Antibody, Novus Biologicals™
SDP

Catalog No. NBP214918 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP214918 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP214918 Supplier Novus Biologicals Supplier No. NBP214918
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRIPT Polyclonal specifically detects CRIPT in Mouse samples. It is validated for Western Blot.

Specifications

Antigen CRIPT
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Accession No. Q9P021
Gene Alias Cysteine-rich interactor of PDZ three, Cysteine-rich interactor of PDZ3, cysteine-rich PDZ-binding protein, HSPC139, postsynaptic protein CRIPT
Gene Symbols CRIPT
Host Species Rabbit
Immunogen Full length human CRIPT Recombinant protein.
Molecular Weight of Antigen 11 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9419
Target Species Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.