Learn More
Cripto-1 TDGF1 Polyclonal Antibody, Rockland™

Rabbit Polyclonal Antibody
$174.73 - $636.24
Specifications
Antigen | Cripto-1 TDGF1 |
---|---|
Concentration | 0.97 mg/mL |
Applications | ELISA, Immunohistochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-1911
![]() ![]() |
Rockland Immunochemicals
600401997 |
100 μg |
Each for $636.24
|
|
|||||
50-199-1912
![]() ![]() |
Rockland Immunochemicals
600401997S |
25 μL |
Each for $174.73
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. This antibody is specific for human Cripto-1 protein. A BLAST analysis was used to suggest limited cross-reactivity with Cripto-1 from mouse based on a 70% homology with the immunizing sequence. Expect cross-reactivity with human Cripto-3 (TDGF2) based on very high levels of sequence conservation. Cross-reactivity with Cripto-1 from other sources has not been determined.
Cripto (TDGF1) is a GPI-anchored cell membrane proetin involved in Nodal signaling. Cell associated TDGF1 acts as a a Nodeal coreceptor in cis. Shedding of TDGF1 by TMEM8A modulated Nodal signaling by allowing soluble TDGF1 to act as a Nodal coreceptor on other cells. TDGF1 is expressed both in ES cells and during the early phases of embryo development, while in adults it is reactivated in a wide range of epithelial cancers.Specifications
Cripto-1 TDGF1 | |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
P13385 | |
6997 | |
Primary | |
Antibody |
0.97 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
CR; CR1; CRGF; CRIPTO; cripto growth factor; Cripto-1; Cripto-1 growth factor; Cripto-3; epidermal growth factor-like Cripto protein; Epidermal growth factor-like cripto protein CR1; TDGF1; TDGF2; teratocarcinoma-derived growth factor; Teratocarcinoma-derived growth factor 1; TERATOCARCINOMA-DERIVED GROWTH FACTOR PRECURSOR (EPIDERMAL GROWTH FACTOR-LIKE CRIPTO PROTEIN) (CRIPTO GROWTH FACTOR) | |
TDGF1 | |
-20°C, Avoid Freeze/Thaw Cycles | |
TDGF1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.