Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIPTO Antibody, Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. N101598A750 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N101598A750 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N101598A750 Supplier Novus Biologicals Supplier No. NB1001598AF750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRIPTO Polyclonal specifically detects CRIPTO in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen CRIPTO
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, teratocarcinoma-derived growth factor 1
Gene Symbols TDGF1
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of mouse Cripto1 (between residues 1-50). [UniProtNo. P51865]
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6997
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.