Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIPTO Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB1001598 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001598 0.1 mL
NB1001598SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB1001598 Supplier Novus Biologicals Supplier No. NB1001598
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

CRIPTO Polyclonal specifically detects CRIPTO in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen CRIPTO
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:1000
Formulation Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
Gene Accession No. P13385
Gene Alias CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, teratocarcinoma-derived growth factor 1
Gene Symbols TDGF1
Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal portion of mouse Cripto1 (between residues 1-50). [UniProt# P51865]
Molecular Weight of Antigen 18 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Neuroscience, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 6997
Target Species Human, Mouse, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.