Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRIPTO Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 3 publications

$192.47 - $529.95

Specifications

Antigen CRIPTO
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/ml, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:1000
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1001598SS
SDP
View Documents
Novus Biologicals
NB1001598SS
0.025 mL
Each for $192.47
Only null left
Add to Cart
 
NB1001598
SDP
View Documents
Novus Biologicals
NB1001598
0.1 mL
Each for $529.95
Only null left
Add to Cart
 
Description

Description

CRIPTO Polyclonal specifically detects CRIPTO in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

CRIPTO
Western Blot 2 ug/ml, Flow Cytometry 1:100, Immunocytochemistry/Immunofluorescence 1:1000
Polyclonal
Rabbit
Apoptosis, Cancer, Cellular Markers, Embryonic Stem Cell Markers, Neuroscience, Stem Cell Markers
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, teratocarcinoma-derived growth factor 1
TDGF1
IgG
Affinity Purified
18 kDa
1.0 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
RUO
Human, Mouse
P13385
6997
A synthetic peptide made to an N-terminal portion of mouse Cripto1 (between residues 1-50). [UniProt# P51865]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.