Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISP-3 Antibody (295208), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB304455 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB304455 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB304455 Supplier Novus Biologicals Supplier No. FAB23972V

Mouse Monoclonal Antibody

CRISP-3 Monoclonal antibody specifically detects CRISP-3 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen CRISP-3
Applications Flow Cytometry
Classification Monoclonal
Clone 295208
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CRISP-3MGC126588, CRS3, cysteine-rich secretory protein 3, cysteine-rich secretory protein-3, dJ442L6.3, SGP28Aeg2, specific granule protein (28 kDa), Specific granule protein of 28 kDa
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRISP-3, Asn21-Tyr245 (Ser106Pro and Ala134Ser), Accession # P54108.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10321
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.