Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ CRISP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
$237.00 - $511.50
Specifications
Antigen | CRISP1 |
---|---|
Concentration | 1 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119066
|
Invitrogen™
PA5119066 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5122066
|
Invitrogen™
PA5122066 |
20 μL |
Each for $237.00
|
|
|||||
Description
CRISP1 Polyclonal Antibody for Western Blot
May have a role in sperm-egg fusion and maturation.Specifications
CRISP1 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Mouse, Rat | |
CRISP1 | |
Acidic epididymal glycoprotein homolog; acidic epididymal glycoprotein-like 1; AEGL1; AEG-like protein; AEG-related protein; ARP; CRISP1; CRISP-1; cysteine rich secretory protein 1; cysteine-rich secretory protein 1; cysteine-rich secretory protein-1 delta; epididymis secretory protein Li 57; HEL-S-57; HSCRISP1D; HSCRISP1G; HUMARP | |
CRISP1 | |
Primary | |
Antigen affinity chromatography, Protein A |
1 mg/mL | |
Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
P12020, Q03401 | |
11571, 654517 | |
Recombinant protein Cysteine Rich Secretory Protein 1. The antigen corresponds to amino acid range 20-244 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title