Learn More
Description
Specifications
Specifications
| Antigen | CRISPR-Cas9 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Clone | 6H4 |
| Conjugate | Alexa Fluor 700 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Cas9, CRISPR, CRISPR-associated protein 9 nuclease |
| Host Species | Mouse |
| Immunogen | Partial recombinant protein made to an N terminal portion of the Staphylococcus aureus CRISPR/Cas9 protein |
| Purification Method | Protein A or G purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
