Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Antibody (7A9-3A3), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP236440IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP236440IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP236440IR Supplier Novus Biologicals Supplier No. NBP236440IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRISPR-Cas9 Monoclonal specifically detects CRISPR-Cas9 in Mouse, Bacteria samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount, Knockout Validated.

Specifications

Antigen CRISPR-Cas9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoblot, Immunoassay
Classification Monoclonal
Clone 7A9-3A3
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount, Knockout Validated
Gene Alias Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Host Species Mouse
Immunogen This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Gene Editing, Gene Knockouts, Gene Therapy
Primary or Secondary Primary
Test Specificity This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus is specific toCas9 protein from Streptococcus pyogene serotype M1.
Target Species Mouse, Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.