Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Antibody (7A9-3A3), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB337040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB337040 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB337040 Supplier Novus Biologicals Supplier No. NBP236440MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRISPR-Cas9 Monoclonal antibody specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot, Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoblot, Immunohistochemistry, KnockDown

Specifications

Antigen CRISPR-Cas9
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone 7A9-3A3
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Immunoblotting, Immunohistochemistry Whole-Mount, Knockout Validated
Formulation 50mM Sodium Borate
Gene Alias Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Host Species Mouse
Immunogen This CRISPR-Cas9 antibody (7A9-3A3) - N-Terminus was raised against Recombinant Cas9 within the N-terminal region of Streptococcus pyogene. [Uniprot: Q99ZW2].
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Gene Editing, Gene Knockouts, Gene Therapy
Primary or Secondary Primary
Gene ID (Entrez) 901176
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.