Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cas9 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NBP379AF488 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP379AF488 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP379AF488 Supplier Novus Biologicals Supplier No. NBP266379AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRISPR-Cas9 Polyclonal specifically detects CRISPR-Cas9 in Bacteria samples. It is validated for Western Blot.

Specifications

Antigen CRISPR-Cas9
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cas9, CRISPR, CRISPR-associated protein 9 nuclease
Host Species Rabbit
Immunogen Full length S. pyogenes Cas9 protein [UniProt Q99ZW2]
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Gene Editing, Gene Knockouts, Gene Therapy
Primary or Secondary Primary
Test Specificity Cas9 protein from Streptococcus pyogene serotype M1.
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.