Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cpf1 Antibody (3D3-F7), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB009557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB009557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB009557 Supplier Novus Biologicals Supplier No. NBP278820AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRISPR-Cpf1 Monoclonal specifically detects CRISPR-Cpf1 in Bacterial samples. It is validated for Western Blot.

Specifications

Antigen CRISPR-Cpf1
Applications Western Blot
Classification Monoclonal
Clone 3D3-F7
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1
Host Species Mouse
Immunogen Partial recombinant protein made to a C-terminal portion of the Acidaminococcus sp. CRISPR-Cpf1 protein
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody is specific for the Acidaminococcus sp. CRISPR-Cpf1
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.