Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRISPR-Cpf1 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP262735C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262735C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP262735C Supplier Novus Biologicals Supplier No. NBP262735C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRISPR-Cpf1 Polyclonal specifically detects CRISPR-Cpf1 in Bacterial samples. It is validated for Western Blot.

Specifications

Antigen CRISPR-Cpf1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Cas12a, Clustered regularly interspaced short palindromic repeat (CRISPR) associated Cas12a, Cpf1, CRISPR-associated endonuclease Cas12a, CRISPR-associated endonuclease Cpf1, CRISPR-Cas12a, FnCas12a, FnCpf1
Host Species Rabbit
Immunogen Partial recombinant Francisella tularensis protein (between amino acids 600-850) [NCBI WP_003034647.1]
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.