Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CrkL Antibody (5E12), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP237586
Description
CrkL Monoclonal specifically detects CrkL in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
CrkL | |
Monoclonal | |
Unconjugated | |
Ascites with 0.03% Sodium Azide | |
CRKL | |
Purified recombinant fragment of human CRKL expressed in E. Coli. | |
Unpurified | |
RUO | |
Primary | |
Human | |
Ascites |
Western Blot, Flow Cytometry, ELISA | |
5000000000000 | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
crk-like protein, pCRKL, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like | |
Mouse | |
39 kDa | |
0.1 mL | |
Cancer, Signal Transduction | |
1399 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction