Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CrkL Antibody (5E12), Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | CrkL |
---|---|
Clone | 5000000000000 |
Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
Applications | Western Blot, Flow Cytometry, ELISA |
Classification | Monoclonal |
Description
CrkL Monoclonal specifically detects CrkL in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
CrkL | |
Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
Monoclonal | |
Ascites | |
RUO | |
Human | |
crk-like protein, pCRKL, v-crk avian sarcoma virus CT10 oncogene homolog-like, v-crk sarcoma virus CT10 oncogene homolog (avian)-like | |
CRKL | |
IgG1 | |
Unpurified |
5000000000000 | |
Western Blot, Flow Cytometry, ELISA | |
Unconjugated | |
Mouse | |
Cancer, Signal Transduction | |
Ascites with 0.03% Sodium Azide | |
1399 | |
Purified recombinant fragment of human CRKL expressed in E. Coli. | |
Primary | |
39 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title