Learn More
Description
Specifications
Specifications
| Antigen | CRM1 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 550 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Chromosome region maintenance 1 protein homolog, DKFZp686B1823, emb, exp1, exportin 1 (CRM1 homolog, yeast), exportin 1 (CRM1, yeast, homolog), exportin-1, Exportin-1 (required for chromosome region maintenance), yeast, homolog |
| Gene Symbols | XPO1 |
| Host Species | Rabbit |
| Immunogen | The immunogen recognized by this antibody maps to a region between residue 1025 and the C-terminus (residue 1071) of human Chromosome Region Maintenance 1 (Exportin 1) (NP_003391.1). |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
