Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRM1 Antibody, Novus Biologicals™
SDP

Catalog No. NB10079811 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10079811 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10079811 Supplier Novus Biologicals Supplier No. NB10079811
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CRM1 Polyclonal specifically detects CRM1 in Human samples. It is validated for Immunoprecipitation, Western Blot, Western Blot (Negative).

Specifications

Antigen CRM1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_003391.1
Gene Alias Chromosome region maintenance 1 protein homolog, DKFZp686B1823, emb, exp1, exportin 1 (CRM1 homolog, yeast), exportin 1 (CRM1, yeast, homolog), exportin-1, Exportin-1 (required for chromosome region maintenance), yeast, homolog
Gene Symbols XPO1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residues 375 and 425 of human Chromosome Region Maintenance 1 (Exportin 1) using the numbering given in entry NP_003391.1 (GeneID 7514).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7514
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with X. laevis, X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, West Indian ocean coelacanth, Duckbill platypus and Northern white-cheeked gibbon.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.