Learn More
CROP/Luc7A Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | CROP/Luc7A |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2380
![]() |
Bethyl Laboratories, Inc
IHC00600T |
10 μL |
Each for $151.31
|
|
|||||
50-157-2379
![]() |
Bethyl Laboratories, Inc
IHC00600 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat, Chicken, Bovine, Dog, Rabbit, Pig, Panda, Orangutan, Monkey, Gorilla, Chimpanzee, Nile tilap
The function of this protein remains unknown.Specifications
CROP/Luc7A | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
cAMP regulatory element-associated protein 1, cisplatin resistance associated overexpressed protein, cisplatin resistance-associated overexpressed protein, cisplatin resistance-associated-overexpressed protein, CRA, CREAP-1, CRE-associated protein, CRE-associated protein 1, CROP, FLJ11063, hLuc7A, LUC7A, LUC7-like 3 (S. cerevisiae), LUC7-like 3 pre-mRNA splicing factor, OA48-18, okadaic acid-inducible phosphoprotein OA48-18 | |
Between 1 and 50 | |
Primary | |
4° C | |
LUC7L3 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
O95232, Q5SUF2 | |
51747, 67684 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.