Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Werner Co. Cross Arm Strap

Cross arm strap with web, O-Ring, and D-ring.

Supplier:  Werner Co. A111006

 View more versions of this product

Catalog No. 17-103-785


Only null left
Add to Cart

Description

Description

Made with durable polyester webbing. Features large O-ring and standard D-ring for pass through.

Specifications

Specifications

72 x 3.9 in.
Secure Structures
72 x 3.9 x 0.4 in.
0.4 in.
2.4 lb.
1.016 cm
D-ring, O-ring
Cross-arm Strap
9.91 cm
3.9 in.
1.09 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.