Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Crotonaldehyde Antibody (2D3.1), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB261922700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261922700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB261922700 Supplier Novus Biologicals Supplier No. NBP261922AF700

Mouse Monoclonal Antibody

Crotonaldehyde Monoclonal specifically detects Crotonaldehyde in Non-species specific samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Crotonaldehyde
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2D3.1
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen Crotonaldehyde modified Keyhole Limpet Hemocyanin (KLH)
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Specific for Crotonaldehyde modified proteins. Does not cross-react with Acrolein, Hexanoyl Lysine, 4-Hydroxy-2-hexenal, 4-Hydroxy nonenal, Malondialdehyde, or Methylglyoxal modified proteins.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.