Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CRTAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | CRTAC1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
CRTAC1 Polyclonal specifically detects CRTAC1 in Human samples. It is validated for Western Blot.Specifications
CRTAC1 | |
Polyclonal | |
Rabbit | |
Q9NQ79 | |
55118 | |
Synthetic peptides corresponding to CRTAC1 (cartilage acidic protein 1) The peptide sequence was selected from the N terminal of CRTAC1)(50ug). Peptide sequence FTAVTNSVLPPDYDSNPTQLNYGVAVTDVDHDGDFEIVVAGYNGPNLVLK. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ASPIC, ASPIC1CEP-68acidic secreted protein in cartilage, cartilage acidic protein 1, CEP68, chondrocyte expressed protein 68 kDa CEP-68,68 kDa chondrocyte-expressed protein, FLJ10320 | |
CRTAC1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title