Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTAM Antibody (210213), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB175297 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB175297 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB175297 Supplier Novus Biologicals Supplier No. FAB16951E
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRTAM Monoclonal specifically detects CRTAM in Human samples. It is validated for CyTOF, Flow Cytometry, Western Blot.

Specifications

Antigen CRTAM
Applications CyTOF, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 210213
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias CD355 antigen, class I MHC restricted T cell associated molecule, class-I MHC-restricted T cell associated molecule, Class-I MHC-restricted T-cell-associated molecule, cytotoxic and regulatory T cell molecule, cytotoxic and regulatory T-cell molecule
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRTAM, Ser18-Ser286, Accession # O95727
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 56253
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.