Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTH-2/GPR44 Antibody (54N1H4), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP219013Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219013Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219013Y Supplier Novus Biologicals Supplier No. NBP219013AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRTH-2/GPR44 Monoclonal specifically detects CRTH-2/GPR44 in Human samples. It is validated for Western Blot.

Specifications

Antigen CRTH-2/GPR44
Applications Western Blot
Classification Monoclonal
Clone 54N1H4
Conjugate Alexa Fluor 647
Dilution Western Blot
Gene Alias CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44
Gene Symbols PTGDR2
Host Species Mouse
Immunogen A recombinant protein containing multiple external domains of GPR44 (CRTH2) was used as immunogen for the antibody. NP_004769
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 11251
Test Specificity GPR44, or G-protein coupled receptor 44, is also designated as CRTH2 and CD294. GPR44 is expressed specifically on CD4+ Th2 cells and type 2 cytotoxic effector cells
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.