Learn More
Description
Specifications
Specifications
| Antigen | CRTH-2/GPR44 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 54N1H4 |
| Conjugate | Alexa Fluor 700 |
| Dilution | Western Blot |
| Gene Alias | CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44 |
| Gene Symbols | PTGDR2 |
| Host Species | Mouse |
| Immunogen | A recombinant protein containing multiple external domains of GPR44 (CRTH2) was used as immunogen for the antibody. NP_004769 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
