Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRTH-2/GPR44 Antibody (BM16), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB160140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB160140 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB160140 Supplier Novus Biologicals Supplier No. NBP318055V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CRTH-2/GPR44 Monoclonal antibody specifically detects CRTH-2/GPR44 in Human samples. It is validated for Flow Cytometry, Immunofluorescence

Specifications

Antigen CRTH-2/GPR44
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone BM16
Conjugate DyLight 405
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, putative G-protein coupled receptor 44
Host Species Rat
Immunogen Human CRTH-2/GPR44 transfected rat cell line TART/B19-12.10
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 11251
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.