Learn More
United Scientific™ Cryo Coder For Skirted Base Vials
Shop All United Scientific Supplies, Inc Products
Click to view available options
Color:
Blue
Green
Red
White
Yellow
Description
- Designed to fit onto the top of our skirted base externally threaded cryogenic vial closures.
- Provide quick visual identification of specific vials for ease in organizing inventory.
- Available in a range of colors.
- Compatible with United P60102 and P60103 skirted base cryo vials.
Specifications
Specifications
| Color | Red |
| Type | Cryo Coder |
| Vial Type | For Use with United Skirted Base Cryo Vials |