Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cryptosporidium Antibody (BEL 0126) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10064526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10064526 0.1 mg
NB017596 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10064526 Supplier Novus Biologicals Supplier No. NB10064526
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

Cryptosporidium Monoclonal specifically detects Cryptosporidium in Protozoa samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin).

Specifications

Antigen Cryptosporidium
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BEL 0126
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry, ELISA 1:2000-1:8000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Gene Alias Cryptosporidiosis
Host Species Mouse
Immunogen Purified oocysts from bovine faeces
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes a membrane antigen expressed by the oocysts of Cryptosporidium sp., an obligate intracellular parasite of the phylum Apicomplexa. The genome of Cryptosporidium sp. is one of the smallest among unicellular eukaryotic organisms. Cryptosporidium is one of several species that cause cryptosporidiosis; a disease affecting the intestines of mammals which is spread through the faecal-oral route. The main symptom is self-limiting diarrhoea in people with intact immune systems. However, in immunocompromised individuals, such as AIDS patients, infection can cause permanent & life-threatening diarrhoea. Cryptosporidiosis is one of the most common waterborne diseases worldwide.
Target Species Protozoa
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.