Learn More
Cryptosporidium Antibody (BEL 0126), Janelia Fluor™ 549, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | Cryptosporidium |
| Applications | ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | BEL 0126 |
| Conjugate | Janelia Fluor 549 |
| Dilution | Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Cryptosporidiosis |
| Host Species | Mouse |
| Immunogen | Purified oocysts from bovine faeces |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.