Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CRYZL1 Antibody (OTI2F9), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N271522PEC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271522PEC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271522PEC5 Supplier Novus Biologicals Supplier No. NBP271522PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CRYZL1 Monoclonal specifically detects CRYZL1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen CRYZL1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI2F9
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias crystallin, zeta (quinone reductase)-like 1, human quinone oxidoreductase homolog-1104P11QOH-1quinone oxidoreductase-like protein 1, Protein 4P11, Quinone oxidoreductase homolog 1, quinone reductase-like 1, Zeta-crystallin homolog
Gene Symbols CRYZL1
Host Species Mouse
Immunogen Full length recombinant protein of human CRYZL1 (NP_665857) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9946.0
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.