Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSDE1 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB291002 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB291002 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB291002 Supplier Novus Biologicals Supplier No. NBP171914APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CSDE1 Polyclonal antibody specifically detects CSDE1 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative)

Specifications

Antigen CSDE1
Applications Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Immunoprecipitation, Western Blot (Negative)
Formulation PBS
Gene Alias cold shock domain containing E1, RNA-binding, cold shock domain-containing protein E1, D1S155EUNRRP5-1000E10.3, DKFZp779B0247, DKFZp779J1455, FLJ26882, KIAA0885, N-ras upstream gene protein, NRAS-related, NRU, Protein UNR, upstream of NRAS
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 600 and 650 of human Upstream of NRAS using the numbering given in entry NP_001007554.1 (GeneID 7812).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7812
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.