Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSK Antibody (7C6), PE, Novus Biologicals™
SDP

Catalog No. NB308803 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB308803 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB308803 Supplier Novus Biologicals Supplier No. NBP259500PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSK Monoclonal antibody specifically detects CSK in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen CSK
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 7C6
Conjugate PE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation PBS
Gene Alias C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK
Host Species Mouse
Immunogen Recombinant human CSK (1-450aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1445
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.