Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSK Antibody (CSK-04), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB500344FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500344FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500344FR Supplier Novus Biologicals Supplier No. NB500344FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSK Monoclonal specifically detects CSK in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen CSK
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CSK-04
Conjugate DyLight 680
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Gene Alias C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK
Gene Symbols CSK
Host Species Mouse
Immunogen Bacterially expressed recombinant fragment of human Csk corresponding to amino acids 330-450.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1445
Test Specificity The antibody CSK-04 reacts with Csk (C-terminal Src kinase), a 50 kDa non-receptor protein tyrosine kinase expressed in cytosol.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.