Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSK Antibody (CSK-04), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB359345 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359345 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359345 Supplier Novus Biologicals Supplier No. NB500344MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CSK Monoclonal antibody specifically detects CSK in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen CSK
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Clone CSK-04
Conjugate mFluor Violet 610 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias C-Src kinase, c-src tyrosine kinase, EC 2.7.10, EC 2.7.10.2, MGC117393, Protein-tyrosine kinase CYL, tyrosine-protein kinase CSK
Host Species Mouse
Immunogen Bacterially expressed recombinant fragment of human Csk corresponding to amino acids 330-450.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 1445
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.