Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSMD1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP229341 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP229341 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP229341 Supplier Novus Biologicals Supplier No. NBP229341

Goat Polyclonal Antibody

CSMD1 Polyclonal specifically detects CSMD1 in Human samples. It is validated for Western Blot, Peptide ELISA.

Specifications

Antigen CSMD1
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:128000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_150094.4
Gene Alias CUB and sushi domain-containing protein 1, CUB and sushi multiple domains protein 1, KIAA1890, UNQ5952/PRO19863
Gene Symbols CSMD1
Host Species Goat
Immunogen Peptide with sequence C-KGVREVNETVTKTP corresponding to internal region according to NP_150094.4.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64478
Test Specificity Reacts with Human. Expected from sequence similarity: Mouse, Rat, Cow.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.