Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CSMD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550645
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PA550645 100 μL
1 options

Catalog No. PA550645

Supplier: Invitrogen™ PA550645

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total CSMD1 protein.

CSMD1 CUB and Sushi multiple domains 1 is a protein that in humans is encoded by the CSMD1gene. It is a potential tumour suppressor, the deletion of which may result in head and neck carcinomas behaving more aggressively. CSMD1 is part of the complement system that defends against pathogens through either the classical pathway or the alternative pathway. Located primarily in nerve growth cones, CSMD1 blocks the classical pathway of the immune system and is thought to be involved in tumor suppression, as defects in the gene encoding CSMD1 are associated with squamous cell carcinomas.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CSMD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CSMD1
Gene Accession No. Q923L3, Q96PZ7
Gene Alias B930082H09; Csmd1; CUB and sushi domain-containing protein 1; CUB and Sushi multiple domains 1; CUB and sushi multiple domains protein 1; KIAA1890; Mdcr1; mKIAA1890; multiple domain complement regulator 1; PPP1R24; protein phosphatase 1, regulatory subunit 24; UNQ5952/PRO19863
Gene Symbols CSMD1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues near the C terminal of human CUB and Sushi multiple domains 1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64478, 94109
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.