Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CSN1S1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$191.80 - $404.50
Specifications
Antigen | CSN1S1 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
FEPA5145435
|
Invitrogen™
PA5145435 |
200 μL |
Each for $404.50
|
|
|||||
FEPA5145436
|
Invitrogen™
PA5145436 |
20 μL |
Each for $191.80
|
|
|||||
Description
CASA play an important role in the capacity of milk to transport calcium phosphate. Casoxin D acts as opioid antagonist and has vasorelaxing activity mediated by bradykinin B1 receptors.Specifications
CSN1S1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P19228, P47710 | |
12990, 1446 | |
Recombinant Human CSN1 protein, Arg16-Trp185 (Accession #P47710). | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
AA959832; alpha S1-casein B; alpha-casein; Alpha-S1-casein; alpha-s1-casein precursor; Antioxidant peptide; AW212882; Bos d 8; CASA; casein alpha s1; casein alpha-S1; casein, alpha; Casoxin-D; Csn1; CSN1S1; Csna | |
CSN1S1 | |
IgG | |
Protein A | |
CSN1S1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title